accessibility menu, dialog, popup

UserName

CAS RN 120728-10-1

CH3H3CCH3OOHNOO

CAS RN 120728-10-1

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylate
Synonyms: 1-[(tert-butoxycarbonyl)amino]cyclobutane-1-carboxylate
Molecular Weight (g/mol): 214.24
Molecular Formula: C10H16NO4
InChi Key: ROVVUKFHORPDSM-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)