CAS RN 1208319-26-9
CAS RN 1208319-26-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oclacitinib, TRC
CAS: 1208319-26-9 Molecular Formula: C15H23N5O2S Molecular Weight (g/mol): 337.44 Synonym: trans-N-Methyl-4-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)cyclohexanemethanesulfonamide,JAKi,PF 03394197 IUPAC Name: N-methyl-1-[4-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclohexyl]methanesulfonamide SMILES: CNS(=O)(=O)C[C@H]1CC[C@@H](CC1)N(C)C2=NC=NC3=C2C=CN3