accessibility menu, dialog, popup

UserName

CAS RN 12145-48-1

PtBrBr

CAS RN 12145-48-1

IUPAC Name: platinum(2+) cycloocta-1,5-diene dibromide
Synonyms: platinum(2+) 1,5-cyclooctadiene dibromide
Molecular Weight (g/mol): 463.08
Molecular Formula: C8H12Br2Pt
InChi Key: HFLCCHAOKSABAA-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Pt++].C1CC=CCCC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results