accessibility menu, dialog, popup

UserName

CAS RN 12154-95-9

COCOFeCOFeCO

CAS RN 12154-95-9

IUPAC Name: bis([(cyclopenta-2,4-dien-1-yl)ferrio]methanone); bis(methanidylidyneoxidanium)
Synonyms: cyclopentadienyliron dicarbonyl dimer
Molecular Weight (g/mol): 353.92
Molecular Formula: C14H10Fe2O4
InChi Key: XJSJEKXJJGHIGW-UHFFFAOYSA-N
SMILES: [C-]#[O+].[C-]#[O+].O=[C-][Fe+]C1C=CC=C1.O=[C-][Fe+]C1C=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)