accessibility menu, dialog, popup

UserName

CAS RN 1216540-18-9

NClOHNHCl

CAS RN 1216540-18-9

IUPAC Name: 5-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenylpentanenitrile hydrochloride
Synonyms: 5-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenylpentanenitrile hydrochloride
Molecular Weight (g/mol): 481.46
Molecular Formula: C28H30Cl2N2O
InChi Key: SSZWNUGWOGONQJ-UHFFFAOYSA-N
SMILES: Cl.OC1(CCN(CCCC(C#N)(C2=CC=CC=C2)C2=CC=CC=C2)CC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results