CAS RN 1216989-88-6
CAS RN 1216989-88-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Disopyramide-d14 Tosylate Salt, TRC
CAS: 1216989-88-6 Molecular Formula: C21 H15 D14 N3 O • C7 H8 O3 S Molecular Weight (g/mol): 525.76 Synonym: Disopyramide-D14 (N,N-diisopropyl-D14) tosylate,Disopyramide-d14 tosylate salt,4-Di(isopropyl-d7)amino-2-phenyl-2-(2-pyridyl)butyramide tosylate,(2RS)-4-[Bis[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-phenyl-2-(pyridin-2-yl)butanamide 4-toluenesulfonate salt IUPAC Name: 4-[bis[1,2,2,2-tetradeuterio-1-(trideuteriomethyl)ethyl]amino]-2-phenyl-2-(2-pyridyl)butanamide;4-methylbenzenesulfonic acid SMILES: [2H]C([2H])([2H])C([2H])(N(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H].Cc3ccc(cc3)S(=O)(=O)O