CAS RN 1217201-89-2
CAS RN 1217201-89-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Zonisamide N,N-Dimethylformimidamide, TRC
CAS: 1217201-89-2 Molecular Formula: C11 H13 N3 O3 S Molecular Weight (g/mol): 267.3 Synonym: N'-(Benzisoxazol-3-ylmethylsulfonyl)-N,N-dimethylformimidamide,Zonisamide N,N-Dimethylformimidamide IUPAC Name: N'-(1,2-benzoxazol-3-ylmethylsulfonyl)-N,N-dimethylmethanimidamide SMILES: CN(C)C=NS(=O)(=O)Cc1noc2ccccc12