accessibility menu, dialog, popup

UserName

CAS RN 1218790-50-1

BOOH3CCH3H3CH3CN(E/Z)

CAS RN 1218790-50-1

IUPAC Name: N-cyclohexyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
Synonyms: N-cyclohexyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
Molecular Weight (g/mol): 313.25
Molecular Formula: C19H28BNO2
InChi Key: BAVKTLABAAKQAH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=NC2CCCCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results