CAS RN 1234480-50-2
IUPAC Name:
5-{[2-ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl]amino}-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms:
5-{[2-ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl]amino}-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol):
474.57
Molecular Formula:
C26H30N6O3
InChi Key:
QAPAJIZPZGWAND-UHFFFAOYSA-N
SMILES:
CCOC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C)C3=N2)C=CC(=C1)N1CCC(O)CC1
1
–
2
of
2
results
1
Tocris Bioscience™ XMD 8-92
CAS: 1234480-50-2 Molecular Formula: C26H30N6O3
2
XMD8-92, MedChemExpress
MedChemExpress XMD8-92 is a potent ERK5 (BMK1)/BRD4 inhibitor with Kds of 80 and 190 nM, respectively. XMD8-92 inhibits DCAMKL2, PLK4 and TNK1 with Kds of 190, 600 and 890 nM, respectively. Anti-cancer activity.