CAS RN 123536-14-1
IUPAC Name:
1-azabicyclo[2.2.2]octan-3-amine dihydrochloride
Synonyms:
1-azabicyclo[2.2.2]octan-3-amine dihydrochloride
Molecular Weight (g/mol):
199.12
Molecular Formula:
C7H16Cl2N2
InChi Key:
STZHBULOYDCZET-UHFFFAOYNA-N
SMILES:
Cl.Cl.NC1CN2CCC1CC2
1
–
2
of
2
results
2
(3R)-Aminoquinuclidine Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.