accessibility menu, dialog, popup

UserName

CAS RN 123784-07-6

NSBr

CAS RN 123784-07-6

IUPAC Name: 2-(5-bromothiophen-2-yl)pyridine
Synonyms: 2-(5-bromothiophen-2-yl)pyridine
Molecular Weight (g/mol): 240.12
Molecular Formula: C9H6BrNS
InChi Key: AKNKWLQVFAYRRO-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results