accessibility menu, dialog, popup

UserName

CAS RN 124-28-7

ClH3CNHCH3CH3

CAS RN 124-28-7

IUPAC Name: dimethyl(octadecyl)azanium chloride
Synonyms: dimethyl stearamine
Molecular Weight (g/mol): 334.03
Molecular Formula: C20H44ClN
InChi Key: PFKRTWCFCOUBHS-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)