accessibility menu, dialog, popup

UserName

FSFFFFFN

CAS RN 1240257-90-2

IUPAC Name: 2-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Synonyms: 2-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Molecular Weight (g/mol): 261.19
Molecular Formula: C8H5F6NS
InChi Key: BISFXPOGLOMAMO-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(CC#N)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results