accessibility menu, dialog, popup

UserName

CAS RN 124215-44-7

BOOH3CCH3H3CH3CBr

CAS RN 124215-44-7

IUPAC Name: 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 248.96
Molecular Formula: C9H18BBrO2
InChi Key: CHQXFJUKMDJWHO-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCCBr)OC1(C)C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)