CAS RN 1246819-07-7
CAS RN 1246819-07-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lapatinib-13C2,15N, TRC
CAS: 1246819-07-7 Molecular Formula: C2713C2H26ClFN315NO4S Molecular Weight (g/mol): 584.04 Synonym: N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine-13C2,15N,GSK 572016-13C2,15N,GW 572016-13C2,15N,GW 572016X-13C2,15N SMILES: FC1=CC(COC(C(Cl)=C2)=CC=C2NC3=NC=NC4=CC=C(C5=CC=C(C[15NH][13CH2][13CH2]S(C)(=O)=O)O5)C=C43)=CC=C1