accessibility menu, dialog, popup

UserName

CAS RN 1257648-78-4

BOOH3CH3CCH3H3CNHNN

CAS RN 1257648-78-4

IUPAC Name: [3-(1H-imidazol-1-yl)propyl]({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine
Synonyms: [3-(imidazol-1-yl)propyl]({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 341.26
Molecular Formula: C19H28BN3O2
InChi Key: OJNUJXDRCCIJCB-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CNCCCN2C=CN=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results