CAS RN 1258226-87-7
CAS RN 1258226-87-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ombitasvir, MedChemExpress
MedChemExpress Ombitasvir is a potent inhibitor of the hepatitis C virus protein NS5A, with EC50s of 0.82 to 19.3 pM against HCV genotypes 1 to 5, and 366 pM against genotype 6a.