accessibility menu, dialog, popup

UserName

A(a)

CAS RN 12627-85-9

IUPAC Name: 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
Synonyms: 4-ethenyl-N,N,N-trimethylanilinium divinylbenzene styrene chloride
Molecular Weight (g/mol): 432.05
Molecular Formula: C29H34ClN
InChi Key: BBQMUEOYPPPODD-UHFFFAOYSA-M
SMILES: [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
Quantity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
Color 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)