accessibility menu, dialog, popup

UserName

CH3H2NClHCl

CAS RN 1264198-68-6

IUPAC Name: 1-(4-chloro-3-methylphenyl)methanamine hydrochloride
Synonyms: 1-(4-chloro-3-methylphenyl)methanamine hydrochloride
Molecular Weight (g/mol): 192.08
Molecular Formula: C8H11Cl2N
InChi Key: WVNIGUUQWGGELM-UHFFFAOYSA-N
SMILES: Cl.CC1=C(Cl)C=CC(CN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results