accessibility menu, dialog, popup

UserName

CH3H3CCH3Cl(E/Z)

CAS RN 126764-17-8

IUPAC Name: 1-chloro-6,6-dimethylhept-2-en-4-yne
Synonyms: 1-chloro-6,6-dimethylhept-2-en-4-yne
Molecular Weight (g/mol): 156.65
Molecular Formula: C9H13Cl
InChi Key: ZIXABMZBMHDFEZ-UHFFFAOYSA-N
SMILES: CC(C)(C)C#CC=CCCl
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)