CAS RN 1268882-43-4
CAS RN 1268882-43-4
IUPAC Name:
8,9-diazatricyclo[4.3.0.0²,⁴]nona-1(6),7-diene-7-carboxylic acid
Synonyms:
8,9-diazatricyclo[4.3.0.0²,⁴]nona-1(6),7-diene-7-carboxylic acid
Molecular Weight (g/mol):
164.16
Molecular Formula:
C8H8N2O2
InChi Key:
CUTZNERBKDMLAP-UHFFFAOYNA-N
SMILES:
OC(=O)C1=NNC2=C1CC1CC21