accessibility menu, dialog, popup

UserName

CH3OHONH2POOHHO(E/Z)

CAS RN 127910-31-0

IUPAC Name: 2-amino-4-methyl-5-phosphonopent-3-enoic acid
Synonyms: 2-amino-4-methyl-5-phosphonopent-3-enoic acid
Molecular Weight (g/mol): 209.14
Molecular Formula: C6H12NO5P
InChi Key: BDYHNCZIGYIOGJ-UHFFFAOYNA-N
SMILES: CC(CP(O)(O)=O)=CC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results