accessibility menu, dialog, popup

UserName

CAS RN 128-39-2

CH3H3CCH3HOH3CH3CCH3

CAS RN 128-39-2

IUPAC Name: 2,6-di-tert-butylphenol
Synonyms: 2,6-di-tert-butylphenol 2,6-di-T-butylphenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: DKCPKDPYUFEZCP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)