CAS RN 129-89-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
[(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt (4-N-Desmethylmetamizole Sodium Salt), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Melaminsulfone Sodium Salt, TRC
CAS: 129-89-5 Molecular Formula: C12 H14 N3 O4 S . Na Molecular Weight (g/mol): 319.31 Synonym: [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt,4-N-Desmethylmetamizole Sodium Salt,Melaminsulfone Sodium Salt,Metamizol Sodium Imp. E (EP) as Sodium Salt IUPAC Name: sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)amino]methanesulfonate SMILES: [Na+].CN1N(C(=O)C(=C1C)NCS(=O)(=O)[O-])c2ccccc2