accessibility menu, dialog, popup

UserName

CAS RN 129604-28-0

ClNFFF

CAS RN 129604-28-0

IUPAC Name: 2-chloro-6-(trifluoromethyl)benzonitrile
Synonyms: 2-chloro-6-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 205.56
Molecular Formula: C8H3ClF3N
InChi Key: DFPYRBFQHBGIKI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C#N)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results