accessibility menu, dialog, popup

UserName

CAS RN 130089-98-4

CH3OHNOOCl

CAS RN 130089-98-4

IUPAC Name: 2-[(4-acetylphenyl)amino]-3-chloro-1,4-dihydronaphthalene-1,4-dione
Synonyms: 2-[(4-acetylphenyl)amino]-3-chloronaphthalene-1,4-dione
Molecular Weight (g/mol): 325.75
Molecular Formula: C18H12ClNO3
InChi Key: LSQZKIQSQHZVQS-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(NC2=C(Cl)C(=O)C3=CC=CC=C3C2=O)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)