accessibility menu, dialog, popup

UserName

NNH2NNH2HClHCl

CAS RN 1301706-74-0

IUPAC Name: 1,2-bis(pyridin-2-yl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(pyridin-2-yl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 287.19
Molecular Formula: C12H16Cl2N4
InChi Key: PLICWJFSNALNJH-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=CC=CC=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results