accessibility menu, dialog, popup

UserName

H3CONH2OHHCl

CAS RN 130198-05-9

IUPAC Name: 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol hydrochloride
Synonyms: 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol hydrochloride
Molecular Weight (g/mol): 285.81
Molecular Formula: C15H24ClNO2
InChi Key: NTKXIDDUCSFBBF-UHFFFAOYNA-N
SMILES: Cl.COC1=CC=C(C=C1)C(CN)C1(O)CCCCC1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Industry Type 
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results