accessibility menu, dialog, popup

UserName

CAS RN 13049-01-9

ClH3COONH3

CAS RN 13049-01-9

IUPAC Name: 2-oxo-2-propoxyethan-1-aminium chloride
Synonyms: 2-oxo-2-propoxyethanaminium chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: YDBMNSABUROTEK-UHFFFAOYSA-N
SMILES: [Cl-].CCCOC(=O)C[NH3+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)