accessibility menu, dialog, popup

UserName

HClH3COONNOCH3

CAS RN 130495-35-1

IUPAC Name: hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole chloride
Synonyms: hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole chloride
Molecular Weight (g/mol): 402.92
Molecular Formula: C22H27ClN2O3
InChi Key: FWLPKVQUECFKSW-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=CC=C(CCCOC(CN2C=CN=C2)C2=CC=C(OC)C=C2)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (1)
  • (4)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)