CAS RN 13138-25-5
IUPAC Name:
[(2E)-but-2-en-1-yl]triphenylphosphanium chloride
Synonyms:
(2E)-but-2-en-1-yltriphenylphosphanium chloride
Molecular Weight (g/mol):
352.84
Molecular Formula:
C22H22ClP
InChi Key:
YYTDJYJBYMQMDI-SQQVDAMQSA-M
SMILES:
[Cl-].C\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(2-Butenyl)triphenylphosphonium chloride, 97%
CAS: 13138-25-5 Molecular Formula: C22H22ClP Molecular Weight (g/mol): 352.84 MDL Number: MFCD00051868 InChI Key: YYTDJYJBYMQMDI-SQQVDAMQSA-M Synonym: 2-butenyl triphenylphosphonium chloride,but-2-en-1-yltriphenylphosphonium chloride,2e-but-2-en-1-yltriphenylphosphanium chloride PubChem CID: 11824356 IUPAC Name: [(E)-but-2-enyl]-triphenylphosphanium;chloride SMILES: [Cl-].C\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1