accessibility menu, dialog, popup

UserName

CAS RN 1314216-34-6

BOONNHH3CH3CH3CH3CCH3

CAS RN 1314216-34-6

IUPAC Name: 2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
Synonyms: 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-1,3-benzodiazole
Molecular Weight (g/mol): 258.13
Molecular Formula: C14H19BN2O2
InChi Key: CPQGLMCLTALRHF-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(C=C2N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results