accessibility menu, dialog, popup

UserName

NFFFOOH

CAS RN 131747-40-5

IUPAC Name: 5-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms: 5-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: RKFDCELCLIZRRH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CN=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results