accessibility menu, dialog, popup

UserName

CAS RN 13194-73-5

CH3BrNH2Br

CAS RN 13194-73-5

IUPAC Name: 3,5-dibromo-4-methylaniline
Synonyms: 3,5-dibromo-4-methylaniline
Molecular Weight (g/mol): 264.95
Molecular Formula: C7H7Br2N
InChi Key: AQZDIKCNODUMNY-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(N)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results