CAS RN 1323966-21-7
CAS RN 1323966-21-7
IUPAC Name:
(4-chloro-2-fluoro-3-methoxyphenyl)methanol
Synonyms:
(4-chloro-2-fluoro-3-methoxyphenyl)methanol
Molecular Weight (g/mol):
190.6
Molecular Formula:
C8H8ClFO2
InChi Key:
ULQDMTWMSOVQRT-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C=CC(CO)=C1F
1
–
1
of
1
results
1
4-Chloro-2-fluoro-3-methoxybenzyl alcohol, 97%, Thermo Scientific™
CAS: 1323966-21-7 Molecular Formula: C8H8ClFO2 Molecular Weight (g/mol): 190.598 MDL Number: MFCD19687181 InChI Key: ULQDMTWMSOVQRT-UHFFFAOYSA-N Synonym: 4-chloro-2-fluoro-3-methoxybenzyl alcohol,4-chloro-2-fluoro-3-methoxyphenyl methanol PubChem CID: 66523606 IUPAC Name: (4-chloro-2-fluoro-3-methoxyphenyl)methanol SMILES: COC1=C(C=CC(=C1F)CO)Cl