accessibility menu, dialog, popup

UserName

CAS RN 132549-43-0

H3CCH3(S)HOONHOOH3CH3CCH3

CAS RN 132549-43-0

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}-5-methylhexanoic acid
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]-5-methylhexanoic acid
Molecular Weight (g/mol): 245.32
Molecular Formula: C12H23NO4
InChi Key: XRVAMBSTOWHUMM-VIFPVBQESA-N
SMILES: CC(C)C[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results