accessibility menu, dialog, popup

UserName

NH2NNOO

CAS RN 13269-19-7

IUPAC Name: 2-nitropyridin-3-amine
Synonyms: 2-nitropyridin-3-amine
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: GZBKVUGZEAJYHH-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results