CAS RN 13269-52-8
IUPAC Name:
(3E)-hex-3-ene
Synonyms:
trans-3-hexene
Molecular Weight (g/mol):
84.16
Molecular Formula:
C6H12
InChi Key:
ZQDPJFUHLCOCRG-AATRIKPKSA-N
SMILES:
CC\C=C\CC
1
–
2
of
2
results
1
trans-3-Hexene, 98%
CAS: 13269-52-8 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.16 InChI Key: ZQDPJFUHLCOCRG-AATRIKPKSA-N Synonym: trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene PubChem CID: 638066 IUPAC Name: (E)-hex-3-ene SMILES: CCC=CCC
2
trans-3-Hexene, 98%
CAS: 13269-52-8 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.162 MDL Number: MFCD00009386 InChI Key: ZQDPJFUHLCOCRG-AATRIKPKSA-N Synonym: trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene PubChem CID: 638066 IUPAC Name: (E)-hex-3-ene SMILES: CCC=CCC