accessibility menu, dialog, popup

UserName

CAS RN 132834-59-4

NNHNFFFCl

CAS RN 132834-59-4

IUPAC Name: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine
Synonyms: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine
Molecular Weight (g/mol): 265.66
Molecular Formula: C10H11ClF3N3
InChi Key: RRIPRMGRQRYRRG-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results