CAS RN 133059-99-1
CAS RN 133059-99-1
IUPAC Name:
hydrogen N-ethyl-1,2-dimethyl-6-(methylimino)-N-phenyl-1,6-dihydropyrimidin-4-amine chloride
Synonyms:
hydrogen N-ethyl-1,2-dimethyl-6-(methylimino)-N-phenylpyrimidin-4-amine chloride
Molecular Weight (g/mol):
292.81
Molecular Formula:
C15H21ClN4
InChi Key:
DUWKUHWHTPRMAP-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CCN(C1=CC=CC=C1)C1=CC(=NC)N(C)C(C)=N1
1
–
2
of
2
results
1
Tocris Bioscience™ ZD 7288
CAS: 133059-99-1 Molecular Formula: C15H21ClN4
2
ZD7288, MedChemExpress
MedChemExpress ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker.