accessibility menu, dialog, popup

UserName

CH3OOHHONOO

CAS RN 134308-13-7

IUPAC Name: 5-(4-methylbenzoyl)-3-nitrobenzene-1,2-diol
Synonyms: tolcapone tasmar
Molecular Weight (g/mol): 273.24
Molecular Formula: C14H11NO5
InChi Key: MIQPIUSUKVNLNT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C1=CC(O)=C(O)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)