accessibility menu, dialog, popup

UserName

CAS RN 13433-00-6

HClH3COONH2OOCH3

CAS RN 13433-00-6

IUPAC Name: hydrogen 1,3-diethyl 2-aminopropanedioate chloride
Synonyms: hydrogen 1,3-diethyl 2-aminopropanedioate chloride
Molecular Weight (g/mol): 211.64
Molecular Formula: C7H14ClNO4
InChi Key: GLFVNTDRBTZJIY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)C(=O)OCC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)