CAS RN 13434-24-7
CAS RN 13434-24-7
IUPAC Name:
manganese(2+) bis(2-ethylhexanoate)
Synonyms:
manganese(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol):
341.35
Molecular Formula:
C16H30MnO4
InChi Key:
FHRAKXJVEOBCBQ-UHFFFAOYNA-L
SMILES:
[Mn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
1
–
1
of
1
results
1
Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn
CAS: 13434-24-7 Molecular Formula: C16H30MnO4 Molecular Weight (g/mol): 341.35 MDL Number: MFCD00058698 InChI Key: FHRAKXJVEOBCBQ-UHFFFAOYSA-L Synonym: manganese bis 2-ethylhexanoate,manganese 2-ethylhexanoate,manganese 2+ bis 2-ethylhexanoate,hexanoic acid, 2-ethyl-, manganese 2+ salt,hexanoic acid, 2-ethyl-, manganese 2+ salt 2:1,hexanoic acid, 2-ethyl-, manganese salt,hexanoic acid, 2-ethyl-, manganese salt 1:?,manganese tallate 2-ethylhexanoate,manganous 2-ethylhexanoate,mmanganese 2-ethylhexanoate PubChem CID: 114521 IUPAC Name: 2-ethylhexanoate;manganese(2+) SMILES: CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Mn+2]