accessibility menu, dialog, popup

UserName

CAS RN 13481-25-9

NNNN

CAS RN 13481-25-9

IUPAC Name: pyrazine-2,3-dicarbonitrile
Synonyms: 2,3-pyrazinedicarbonitrile
Molecular Weight (g/mol): 130.11
Molecular Formula: C6H2N4
InChi Key: OTVZGAXESBAAQQ-UHFFFAOYSA-N
SMILES: N#CC1=NC=CN=C1C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results