CAS RN 134993-88-7
IUPAC Name:
(3,5-difluorophenyl)hydrazine hydrochloride
Synonyms:
(3,5-difluorophenyl)hydrazine hydrochloride
Molecular Weight (g/mol):
180.58
Molecular Formula:
C6H7ClF2N2
InChi Key:
CKCNKYAHVKNKHQ-UHFFFAOYSA-N
SMILES:
Cl.NNC1=CC(F)=CC(F)=C1
1
–
1
of
1
results
1
3,5-Difluorophenylhydrazine hydrochloride, 97+%, Thermo Scientific™
CAS: 134993-88-7 Molecular Formula: C6H6F2N2·HCl Molecular Weight (g/mol): 180.58 MDL Number: MFCD03094171 InChI Key: CKCNKYAHVKNKHQ-UHFFFAOYSA-N Synonym: 3,5-difluorophenylhydrazine hydrochloride,3,5-difluorophenyl hydrazine hydrochloride,3,5-difluorophenylhydrazine hcl,3,5-difluorophenylhydrazinehydrochloride,1-3,5-difluorophenyl hydrazine hydrochloride,3,5-difluoro phenylhydrazine,hydrazine, 3,5-difluorophenyl-, monohydrochloride,pubchem1085,pubchem20776,c6h6f2n2.hcl PubChem CID: 16217172 IUPAC Name: (3,5-difluorophenyl)hydrazine;hydrochloride SMILES: C1=C(C=C(C=C1F)F)NN.Cl