CAS RN 136-44-7
CAS RN 136-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lisadimate (>85%), TRC
CAS: 136-44-7 Molecular Formula: C10H13NO4 Molecular Weight (g/mol): 211.21 Synonym: Glyceryl p-aminobenzoate,Escalol 106,Glycerol, 1-(p-aminobenzoate),2,3-Dihydroxypropyl 4-aminobenzoate,4-Aminobenzoic acid 1-glyceryl ester,NIPA GMPA IUPAC Name: 2,3-dihydroxypropyl 4-aminobenzoate SMILES: Nc1ccc(cc1)C(=O)OCC(O)CO