accessibility menu, dialog, popup

UserName

CAS RN 136-53-8

ZnH3CCH3OOH3CCH3OO

CAS RN 136-53-8

IUPAC Name: zinc(2+) bis(2-ethylhexanoate)
Synonyms: zinc ethylhexanoate
Molecular Weight (g/mol): 351.79
Molecular Formula: C16H30O4Zn
InChi Key: IFNXAMCERSVZCV-UHFFFAOYNA-L
SMILES: [Zn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results