accessibility menu, dialog, popup

UserName

CAS RN 136632-32-1

H3CCH3ONHH3CH3COHNSH3COHNOCH3

CAS RN 136632-32-1

IUPAC Name: 2-acetamido-4-methyl-N-(3-methyl-1-{[4-(methylsulfanyl)-1-oxobutan-2-yl]carbamoyl}butyl)pentanamide
Synonyms: 2-acetamido-4-methyl-N-(3-methyl-1-{[4-(methylsulfanyl)-1-oxobutan-2-yl]carbamoyl}butyl)pentanamide
Molecular Weight (g/mol): 401.57
Molecular Formula: C19H35N3O4S
InChi Key: RJWLAIMXRBDUMH-UHFFFAOYNA-N
SMILES: CSCCC(NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(C)=O)C=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results