CAS RN 136790-76-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Lubiprostone, TRC
CAS: 136790-76-6 Molecular Formula: C20 H32 F2 O5 Molecular Weight (g/mol): 390.46 Synonym: (11α)-16,16-difluoro-11-hydroxy-9,15-dioxo-Prostan-1-oic acid,Lubiprostone,Ru 0211 IUPAC Name: 7-[(1R,2R,3R)-2-(4,4-difluoro-3-oxooctyl)-3-hydroxy-5-oxocyclopentyl]heptanoic acid SMILES: CCCCC(F)(F)C(=O)CC[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)O
2
Lubiprostone, MedChemExpress
MedChemExpress Lubiprostone(SPI-0211;RU0211) is a gastrointestinal agent used for the treatment of idiopathic chronic constipation.