CAS RN 13720-08-6
CAS RN 13720-08-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4-Di(methyl-d3)-naphthalene, TRC
CAS: 13720-08-6 Molecular Formula: C12H6D6 Molecular Weight (g/mol): 162.26 Synonym: NSC 61779-d6,(1,4-Dimethyl-d6)-naphthalene SMILES: [2H]C([2H])([2H])c1ccc(c2ccccc12)C([2H])([2H])[2H]